Wasser, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
4217.9020EA
0
EUR
4217.9020
Wasser, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Wasser
Formel:
H₂O Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Ammonium (NH4) | max. 0.3 ppm |
Conductivity (µS/cm) | max. 5 |
Conductivity at 20°C | max. 5 |
Heavy Metals (as Pb) | passes test |
pH at 25°C | 5.0 - 7.0 |
Residue after Evaporation | max. 10 ppm |
Substances Reducing KMnO4 | passes test |
Trace Impurities | |
Chloride (Cl) | max. 1 ppm |
Sulfate (SO4) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...