Wasser, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®

Lieferant: Avantor

Synonyme: Wasserstoffoxid, Hydrogenoxid

M.O.C. Category NI

4218.2500EA 49.3 EUR
4218.2500 4218.1000
Wasser, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®
Wasser
Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Particulate Matter max. 0.0001 %
Residue after Evaporation max. 2 ppm
Ultraviolet absorbance of largest eluted peak
at 220 nm max. 0.005 au
at 254 nm max. 0.001 au
Physical Data (not specifications):


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR