(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
42183.03
42183.03EA
55.1
EUR
42183.03
(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol ≥97%
(1S,2S)-(+)-2-Amino-1-phenyl-1,3-propanediol
Purity: 98+%
CAS No.: 28143-91-1
Molecular Formula: C9H13NO2
Formula Weight: 167.21
Physical Form: Powder
Melting Point: 112-115°
MDL No.: MFCD00004503
Applications: Asymetric synthesis of chiral 2-oxazolines
Formel:
C₉H₁₃NO₂ Molecular Weight: 167.208 g/mol Schmelzpunkt: 112…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004503 CAS-Nummer: 28143-91-1 EINECS: 248-867-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...