Bis(triphenylphosphin)nickel(II)bromid ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

42192.06
42192.06EA 69.1 EUR
42192.06
Bis(triphenylphosphin)nickel(II)bromid ≥99%
Bis(triphenylphosphin)nickel(II)bromid

Purity: 99% CAS No.: 14126-37-5 Molecular Formula: Br2Ni[P(C6H5)3]2 Formula Weight: 743.12 Physical Form: Powder Odour: Odourless MDL No.: MFCD00015865


As a polymerization catalyst

Formel: C₃₆H₃₀Br₂NiP₂
Schmelzpunkt: 219…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015865
CAS-Nummer: 14126-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR