Polyethylene oxide, M.W. 100000
Lieferant: Thermo Fisher Scientific
Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.
Tg=-67° High viscosity, combustible. Keep away from heat, sparks and open flame. Incompatible with strong acids, strong bases and strong oxidizing agents.
Soluble in toluene, THF, hot water, DMF, CHCl3
Formel:
(-CH₂CH₂O-)n Schmelzpunkt: 65 °C Dichte: 1,4539 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00147970 CAS-Nummer: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...