Polyethylene oxide, M.W. 100000

Lieferant: Thermo Fisher Scientific

42236.14 42236.22 42236.36
42236.14EA 38.1 EUR
42236.14 42236.22 42236.36
Polyethylene oxide, M.W. 100000
Polyethylene oxide

Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.


Tg=-67° High viscosity, combustible. Keep away from heat, sparks and open flame. Incompatible with strong acids, strong bases and strong oxidizing agents.
Soluble in toluene, THF, hot water, DMF, CHCl3

Formel: (-CH₂CH₂O-)n
Schmelzpunkt: 65 °C
Dichte: 1,4539
Lagertemperatur: Raumtemperatur
MDL: MFCD00147970
CAS-Nummer: 25322-68-3
EINECS: 500-038-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR