Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Folie, Dicke 0.2 mm (0.008 in) Type 304
Lieferant: Thermo Scientific
|
Gefahr
|
42255.GH
42255.GHEA
70.1
EUR
42255.GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Folie, Dicke 0.2 mm (0.008 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odour: Odourless
MDL No.: MFCD00148499