Stirring Rods, Thermo Fisher Scientific
Lieferant: Thermo Fisher Scientific
42294.NK 42293.NK 42292.NT 42293.NT 42294.NT
42294.NKEA 27.1 EUR
42294.NK 42293.NK 42292.NT 42293.NT 42294.NT
Stirring Rods, Thermo Fisher Scientific
Rührer Rührstäbe
The fused quartz rods are manufactured by fusing naturally occurring crystalline silica.
  • Fused quartz has high chemical purity, high corrosion resistance and high melting point
  • Quartz is extremely hard, with a low coefficient of thermal expansion, excellent electrical insulation qualities and optical transmission from ultra-violet to infra-red