2-Ethylhexylmethacrylat ≥98% stabilisiert
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Stabilisierung:
Stabilisiert mit Hydrochinonmonomethylether (4-Methoxyphenol, MEHQ)
Stabilized with 4-methoxyphenol
Purity: 98%
CAS No.: 688-84-6
Molecular Formula: C12H22O2
Formula Weight: 198.31
Flash Point: 92°(198°F)
Physical Form: Liquid
Density: 0.885
Refractive Index: 1.4380
Appearance: Colourless
Odour: Ether-like
Melting Point: -50°
Boiling Point: 218°
Beilstein Reference: 1769420
MDL No.: MFCD00009494
Monomer used in the preparation of polymethacrylate polymers
Formel:
C₁₂H₂₂O₂ Siedepunkt: 218 °C Schmelzpunkt: –50 °C Dichte: 0,885 Flammpunkt: 92 °C (198 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009494 CAS-Nummer: 688-84-6 EINECS: 211-708-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...