Gold-Beschichtungslösung ∼3,7 g/l (Metalgehalt), stromlos
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42307.AK
42307.K2
42307.AKEA
271
EUR
42307.AK
42307.K2
Gold-Beschichtungslösung ∼3,7 g/l (Metalgehalt), stromlos
Gold plating solution
Physical Form: Liquid
Appearance: Colourless to pale yellow
UN No.: UN2922
MDL No.: MFCD00003436
Applications: For coating nickel or copper
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00003436 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...