Tantal ≥99,95% (Metall-Basis), Stäbe, Ø 2.8 mm (0.11 in)
Lieferant: Thermo Fisher Scientific
42371.BU
42371.BY
42371.BUEA
214
EUR
42371.BU
42371.BY
Tantal ≥99,95% (Metall-Basis), Stäbe, Ø 2.8 mm (0.11 in)
Tantalum
Purity: 99.95%
CAS No.: 7440-25-7
Physical Form: ≈102g/m
Odour: Odourless
MDL No.: MFCD00011252
Formel:
Ta Molecular Weight: 180.95 g/mol Siedepunkt: 5427 Schmelzpunkt: 2996 Dichte: 16.69 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011252 CAS-Nummer: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...