(3-Chlorpropyl)trimethoxysilan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
Packaged under Argon in resealable ChemSeal® bottles
Purity: 97+%
CAS No.: 2530-87-2
Molecular Formula: ClCH2CH2CH2Si(OCH3)3
Formula Weight: 198.72
Flash Point: 78°(172°F)
Physical Form: Liquid
Density: 1.077
Refractive Index: 1.4200
Sensitivity: Moisture Sensitive
Odour: Recognizable
Melting Point: -50°
Boiling Point: 195-197°
Beilstein Reference: 969627
MDL No.: MFCD00000997
Formel:
ClH₂CCH₂CH₂Si(OCH₃)₃ Siedepunkt: 195…197 °C Schmelzpunkt: –50 °C Dichte: 1,077 Flammpunkt: 78 °C (172 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000997 CAS-Nummer: 2530-87-2 EINECS: 219-787-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...