Methoxytrimethylsilan ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Trimethyl(methoxy)silan
42464.09
42464.18
42464.30
42464.09EA
35.7
EUR
42464.09
42464.18
42464.30
Methoxytrimethylsilan ≥97%
Methoxytrimethylsilan
Purity: 97+%
CAS No.: 1825-61-2
Molecular Formula: C4H12OSi
Formula Weight: 104.22
Flash Point: -11°(-12°F)
Physical Form: Liquid
Density: 0.756
Refractive Index: 1.3678
Sensitivity: Moisture Sensitive
Odour: Mild
Boiling Point: 57-58°
UN No.: 1993
MDL No.: MFCD00008344
Formel:
C₄H₁₂OSi Siedepunkt: 57…58 °C Dichte: 0,756 Flammpunkt: –11 °C (–12 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008344 CAS-Nummer: 1825-61-2 EINECS: 217-369-0 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...