Silber ≥99,998% (Metall-Basis), Drahtform, Premion®, Ø 1.5 mm (0.06 in)

Lieferant: Thermo Fisher Scientific

42487.BU 42487.BY 42487.CC
42487.BUEA 87 EUR
42487.BU 42487.BY 42487.CC
Silber ≥99,998% (Metall-Basis), Drahtform, Premion®, Ø 1.5 mm (0.06 in)
Silver

Purity: 99.998% CAS No.: 7440-22-4 Physical Form: ≈18.54g/m Odour: Characteristic MDL No.: MFCD00003397

Formel: Ag
Siedepunkt: 2212 °C (1013 hPa)
Schmelzpunkt: 962 °C
Dichte: 10,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003397
CAS-Nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR