Ion chromatography standard solution, acetate, Specpure®, 1000 µg/ml

Lieferant: Thermo Fisher Scientific

42563.AE 42563.AP
42563.AEEA 141 EUR
42563.AE 42563.AP
Ion chromatography standard solution, acetate, Specpure®, 1000 µg/ml
Standards Anionische Standards Acetat-Standards

Molecular Formula: CH₃CO₂Na in H₂O
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00282007

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
MDL No. MFCD00282007
Molecular formula CH3CO2Na in H2O
Physical form Liquid
Odour/scent Odourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR