Chromate, ion chromatography standard solution, Specpure®, CrO₄²⁻ 1000 µg/ml
Lieferant: Thermo Fisher Scientific
42564.AE
42564.AP
42564.AEEA
125
EUR
42564.AE
42564.AP
Chromate, ion chromatography standard solution, Specpure®, CrO₄²⁻ 1000 µg/ml
Standards
Anionische Standards
Chromat (VI)-Standards
Molecular Formula: K₂CrO₄ in H₂O
Physical Form: Liquid
Appearance: Yellow
Odour: Odourless
MDL No.: MFCD00011368
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...