Chromate, ion chromatography standard solution, Specpure®, CrO₄²⁻ 1000 µg/ml

Lieferant: Thermo Fisher Scientific

42564.AE 42564.AP
42564.AEEA 125 EUR
42564.AE 42564.AP
Chromate, ion chromatography standard solution, Specpure®, CrO₄²⁻ 1000 µg/ml
Standards Anionische Standards Chromat (VI)-Standards

Molecular Formula: K₂CrO₄ in H₂O
Physical Form: Liquid
Appearance: Yellow
Odour: Odourless
MDL No.: MFCD00011368

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR