Formate, Ion chromatography standard solution, Specpure®, HCO₂- 1000 µg/ml

Lieferant: Thermo Fisher Scientific

42566.AE 42566.AP
42566.AEEA 147 EUR
42566.AE 42566.AP
Formate, Ion chromatography standard solution, Specpure®, HCO₂- 1000 µg/ml
Standards Anionische Standards Formate-Standards

Molecular Formula: HCO₂Na in H₂O
Physical Form: Liquid
MDL No.: MFCD00003297

Bestellen

TECHNISCHE DATEN

Environmentally Preferable
MDL No. MFCD00003297
Molecular formula HCO2Na in H2O
Physical form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR