Oxalate, Ion chromatography standard solution, Specpure®, C_2O4-2 1000 µg/ml
Lieferant: Thermo Fisher Scientific
42568.AE
42568.AEEA
129
EUR
42568.AE
Oxalate, Ion chromatography standard solution, Specpure®, C_2O4-2 1000 µg/ml
Standards
Anionische Standards
Oxalat-Standards
Molecular Formula: Na₂C₂O₄ in H₂O
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00012465
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...