Supelco® Organic Reference Standard, Ethyl 4-(Dimethylamino)Benzoate
Lieferant: Merck
42582-1G
42582-1GEA 137 EUR
42582-1G
Supelco® Organic Reference Standard, Ethyl 4-(Dimethylamino)Benzoate
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P202 - P273 - P280 - P308 + P313 - P391 - P405
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c0a55edb12838c666dc4b355c613c833193eba1&vid=3c0a55edb12838c666dc4b355c613c833193eba1&prid=web&clid=SAPDAM