ICP-MS soil spike standard solution for 6020 CLP-M, Specpure®

Lieferant: Thermo Fisher Scientific

42604.AE
42604.AEEA 289 EUR
42604.AE
ICP-MS soil spike standard solution for 6020 CLP-M, Specpure®
Standards Kationischer Standards Mehrelement-Standards

Molecular Formula: Matrix: 2% HNO₃/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD02266211

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR