Tebbe Reagent 0.5 M in Toluol, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: µ-Chloro-µ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, Bis(cyclopentadienyl-mu-chloro(dimethylaluminium-mu-methylenetitanium

426061000
426061000.EA 517 EUR
426061000.
Tebbe Reagent 0.5 M in Toluol, AcroSeal®
Tebbe Reagent

Appearance: Dark red to purple Solution

Formel: C₁₃H₁₈AlClTi
Molecular Weight: 284,62 g/mol
Dichte: 0,927 g/cm³
Flammpunkt: 4 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00151575
CAS-Nummer: 67719-69-1
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Dark red to purple Solution
Strength 0.40 to 0.60 M
Refractive index 1.4165 to 1.4195 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR