Germanium(IV)isopropoxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
42673.03
42673.06
42673.14
42673.03EA
69.1
EUR
42673.03
42673.06
42673.14
Germanium(IV)isopropoxid ≥99,9% (Metall-Basis)
Germanium(IV)isopropoxid
Purity: 99.9%
CAS No.: 21154-48-3
Molecular Formula: C12H28GeO4
Formula Weight: 308.94
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.025
Refractive Index: 1.414
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Alcohol-like
Boiling Point: 109°/30mm
MDL No.: MFCD00015339
Formel:
C₁₂H₂₈GeO₄ Siedepunkt: 109 °C (30 mmHg) Dichte: 1,025 Flammpunkt: 68 °C (154 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015339 CAS-Nummer: 21154-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...