Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in wässriger Lösung

Lieferant: Thermo Fisher Scientific
Achtung

42676.A1 42676.A7 42676.30
42676.A1EA 220 EUR
42676.A1 42676.A7 42676.30
Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in wässriger Lösung
Dihydroxybis(ammonium lactato)titanium(IV)

CAS No.: 65104-06-5 Molecular Formula: C6H18N2O8Ti Formula Weight: 294.10 Flash Point: 47°(118°F) Physical Form: Liquid Density: 1.21 Refractive Index: 1.4380 Appearance: Pale yellow UN No.: UN1993 MDL No.: MFCD00072736


Applications: Water soluble titanium complex

Formel: C₆H₂₂N₂O₈Ti
Dichte: 1,21
Flammpunkt: 47 °C (118 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00072736
CAS-Nummer: 65104-06-5
EINECS: 247-428-6
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR