Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Achtung
|
42676.A1
42676.A7
42676.30
42676.A1EA
220
EUR
42676.A1
42676.A7
42676.30
Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in wässriger Lösung
Dihydroxybis(ammonium lactato)titanium(IV)
CAS No.: 65104-06-5
Molecular Formula: C6H18N2O8Ti
Formula Weight: 294.10
Flash Point: 47°(118°F)
Physical Form: Liquid
Density: 1.21
Refractive Index: 1.4380
Appearance: Pale yellow
UN No.: UN1993
MDL No.: MFCD00072736
Applications: Water soluble titanium complex
Formel:
C₆H₂₂N₂O₈Ti Dichte: 1,21 Flammpunkt: 47 °C (118 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00072736 CAS-Nummer: 65104-06-5 EINECS: 247-428-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...