p-Tolylmagnesium bromide ∼0.5 M in Diethylether, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
426761000
426768000
426761000.EA
55.9
EUR
426761000.
426768000.
p-Tolylmagnesium bromide ∼0.5 M in Diethylether, AcroSeal®
p-Tolylmagnesium bromide
Formel:
C₇H₇BrMg Dichte: 0,866 g/cm³ Flammpunkt: –40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000040 CAS-Nummer: 4294-57-9 UN: 2924 ADR: 3,I |
Spezifikation Testergebnisse
Appearance | Clear to hazy yellow to amber Solution (May develop some turbidity or precipitate) |
Titration Grignard | 0.40 to 0.60 M |
Rhodium (Rh) | ≥34.7 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...