p-Tolylmagnesium bromide ∼0.5 M in Diethylether, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

426761000 426768000
426761000.EA 55.9 EUR
426761000. 426768000.
p-Tolylmagnesium bromide ∼0.5 M in Diethylether, AcroSeal®
p-Tolylmagnesium bromide
Formel: C₇H₇BrMg
Dichte: 0,866 g/cm³
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000040
CAS-Nummer: 4294-57-9
UN: 2924
ADR: 3,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear to hazy yellow to amber Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M
Rhodium (Rh) ≥34.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR