Isopropylmagnesiumbromid 3 M in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

426781000
426781000.EA 132 EUR
426781000.
Isopropylmagnesiumbromid 3 M in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®
Isopropylmagnesium bromide

Appearance: Clear dark grey to dark brown Solution

Formel: C₃H₇BrMg
Dichte: 1,08 g/cm³
Flammpunkt: –11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672110
CAS-Nummer: 920-39-8
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear dark grey to dark brown Solution
Titration Grignard 2.85 to 3.20
GC ≥99.6 % (SOCl2)
Color scale ≤50 APHA
Impurity ≤0.05 % (SCl2 + S2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR