4-Fluorobenzylmagnesium chloride 0.25 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
426790500
426790500.EA
249
EUR
426790500.
4-Fluorobenzylmagnesium chloride 0.25 M in THF, AcroSeal®
4-Fluorobenzylmagnesium chloride
Appearance: Yellow to orange to brown Solution
Formel:
C₇H₆ClFMg Dichte: 0,948 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319898 CAS-Nummer: 1643-73-8 UN: 2056 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Yellow to orange to brown Solution |
Titration Grignard | 0.15 to 0.35 M |
Refractive index | 1.4690 to 1.4730 (20°C, 589 nm) |
Tin (Sn) | 17.5 to 19.9 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...