Supelco® Organic Reference Standard, 2-Phenylethylamine Hydrochloride
Lieferant: Merck
42681-50MG
42681-50MGEA
86.4
EUR
42681-50MG
Supelco® Organic Reference Standard, 2-Phenylethylamine Hydrochloride
Standards
Organische Standards
- GHS signal: GHS07
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338