Titan(IV)oxid rutil ≥99,8% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

42681.30 42681.A1 42681.A7
42681.30EA 121 EUR
42681.30 42681.A1 42681.A7
Titan(IV)oxid rutil ≥99,8% (Metall-Basis)
Titan(IV)oxid rutil

Purity: 99.8% CAS No.: 1317-80-2 Molecular Formula: TiO2, 97% min rutile phase Formula Weight: 79.90 Physical Form: 0.9-1.6 Micron APS Powder, S.A. 3.3-6.1 m2/g Density: 4.23 Melting Point: 1830-1850° Boiling Point: 2500-3000° Merck Reference: 14,9472 MDL No.: MFCD00011269

Formel: O₂Ti
Siedepunkt: 2500…3000 °C
Schmelzpunkt: 1830…1850 °C
Dichte: 4,23
Lagertemperatur: Raumtemperatur
MDL: MFCD00011269
CAS-Nummer: 1317-80-2
EINECS: 215-282-2
Merck Index: 14,9472

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR