Poly(styrene) 2,5% (w/w) Dispersion in Wasser, Latex-Mikrosphären 0.10 micron

Lieferant: Thermo Fisher Scientific

42712.4Y
42712.4YEA 475 EUR
42712.4Y
Poly(styrene) 2,5% (w/w) Dispersion in Wasser, Latex-Mikrosphären 0.10 micron
Poly(styrene)

CAS No.: 9003-53-6 Molecular Formula: [-CH2CH(C6H5)-]n Storage Temperature: 2°C to 8°C Physical Form: Dispersion Appearance: Opalescent Odour: Odourless MDL No.: MFCD00084450

Formel: C₈H₈
Lagertemperatur: Kühlschrank
MDL: MFCD00084450
CAS-Nummer: 9003-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR