Boran-Pyridin-Komplex ∼9.4 M 95%, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Pyridine Borane, Borane, pyridine complex

427131000
427131000.EA 195 EUR
427131000.
Boran-Pyridin-Komplex ∼9.4 M 95%, AcroSeal®
Boranepyridine complex

Appearance: Clear to slightly hazy colorless to amber Liquid

Formel: C₅H₈BN
Molecular Weight: 92,94 g/mol
Schmelzpunkt: 10…11 °C
Dichte: 0,92 g/cm³
Flammpunkt: 73 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012435
CAS-Nummer: 110-51-0
UN: 2810
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear to slightly hazy colorless to amber Liquid (May darken on storage)
Infrared spectrum Conforms
Titration ≥94 % (Active hydrogen)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR