Propionaldehyd 99+%, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Propanal
427211000
427211000.EA
23.9
EUR
427211000.
Propionaldehyd 99+%, AcroSeal®
Propionaldehyd
Formel:
C₃H₆O Siedepunkt: 48 °C (1013 hPa) Schmelzpunkt: –81 °C Dichte: 0,798 g/cm³ Flammpunkt: –40 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007020 CAS-Nummer: 123-38-6 UN: 1275 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3610 to 1.3660 (20°C, 589 nm) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Acidity/alkalinity | ≤0.00008 meq/g |
Substance darkened by H2SO4 | passes test |
Calcium (Ca) | ≤0.2 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Potassium (K) | ≤0.1 ppm |
Sodium (Na) | ≤0.1 ppm |
Zinc (Zn) | ≤0.1 ppm |
Hydrocarbons | ≤0.1 % Aromatic hydrocarbons |
Total phosphorus (P) | ≤0.1 ppm |
Total silicon (Si) | ≤0.05 ppm |
Total sulfur (S) | ≤10 ppm |
Bromine number | ≤0.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...