Kieselgel, Porengröße: 150 Å, großporig
Lieferant: Thermo Fisher Scientific
|
Achtung
|
42726.18
42726.30
42726.A1
42726.A7
42726.18EA
31.6
EUR
42726.18
42726.30
42726.A1
42726.A7
Kieselgel, Porengröße: 150 Å, großporig
Silica gel
CAS No.: 63231-67-4
Physical Form: -100+200 Mesh, S.A. 350-400m2/g
Odour: Odourless
MDL No.: MFCD00011232
Column chromatography without applied pressure (gravity pulled)
Formel:
SiO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...