Chlor(diisopropyl)phosphin 96%, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Di-i-propylchlorophosphine

427270500
427270500.EA 358 EUR
427270500.
Chlor(diisopropyl)phosphin 96%, AcroSeal®
Chlor(diisopropyl)phosphin
Formel: C₆H₁₄ClP
Molecular Weight: 152,6 g/mol
Siedepunkt: 69 °C (33 mmHg)
Flammpunkt: 3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015027
CAS-Nummer: 40244-90-4
UN: 2924
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
GC ≥95 %
Residue after evaporation ≤0.002 %
Acidity/alkalinity ≤0.0001 meq/g
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm
Total phosphorus (P) ≤0.5 ppm
Total silicon (Si) ≤0.02 ppm
Total sulfur (S) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR