Samarium cobalt (33% Sm), REacton®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42732.18
42732.30
42732.18EA
92.5
EUR
42732.18
42732.30
Samarium cobalt (33% Sm), REacton®
Samarium cobalt
CAS No.: 12017-68-4
Molecular Formula: SmCo5, Intermetallic, Magnet Alloy Additive
Physical Form: Fine Powder
Sensitivity: Air and Moisture Sensitive
Odour: Odourless
UN No.: UN3089
MDL No.: MFCD02091682
Formel:
CoSm Lagertemperatur: Raumtemperatur |
MDL:
MFCD02091682 CAS-Nummer: 12017-68-4 EINECS: 234-625-7 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...