3,5-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
427441000
427441000.EA
139
EUR
427441000.
3,5-Difluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
3,5-Difluorophenylmagnesium bromide
Appearance: Yellow to brown Solution
Formel:
C₆H₃BrF₂Mg Siedepunkt: 65 °C (760 mmHg) Dichte: 0,971 g/cm³ Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672005 CAS-Nummer: 62351-47-7 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Yellow to green or brown Solution |
Titration Grignard | 0.40 to 0.75 M |
Refractive index | 1.5610 to 1.5630 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...