Cobalt(II)carbonat ≥99,5% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42745.18
42745.30
42745.18EA
85
EUR
42745.18
42745.30
Cobalt(II)carbonat ≥99,5% (Metall-Basis)
Cobalt(II) carbonate
Purity: 99.5%
CAS No.: 513-79-1
Molecular Formula: CoCO3
Formula Weight: 118.94
Physical Form: -325 Mesh Powder
Density: 4.13
Melting Point: dec.
Merck Reference: 14,2436
UN No.: 3077
MDL No.: MFCD00010940
Formel:
CCoO₃ Dichte: 4,13 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010940 CAS-Nummer: 513-79-1 EINECS: 208-169-4 UN: 3077 ADR: 9,III Merck Index: 14,2436 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...