o-Tolylmagnesium Bromide 2 M in Diethylether, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

427471000
427471000.EA 139 EUR
427471000.
o-Tolylmagnesium Bromide 2 M in Diethylether, AcroSeal®
o-Tolylmagnesium Bromide

Appearance: Grey to brown Solution

Formel: C₇H₇BrMg
Dichte: 1,013 g/cm³
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010350
CAS-Nummer: 932-31-0
UN: 2924
ADR: 3,I

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown Solution
Titration Active base 1.90 to 2.20 M
Melting point 184 to 187 °C
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR