Poly(styrene) 2,5% (w/w) Dispersion in Wasser, Latex-Mikrosphären 45.0 micron
Lieferant: Thermo Fisher Scientific
42749.AA
42749.AAEA
571
EUR
42749.AA
Poly(styrene) 2,5% (w/w) Dispersion in Wasser, Latex-Mikrosphären 45.0 micron
Poly(styrene)
CAS No.: 9003-53-6
Molecular Formula: [-CH2CH(C6H5)-]n
Storage Temperature: 2°C to 8°C
Physical Form: Dispersion
Appearance: Opalescent
Odour: Odourless
MDL No.: MFCD00084450
Formel:
C₈H₈ Lagertemperatur: Kühlschrank |
MDL:
MFCD00084450 CAS-Nummer: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...