Tetrakis(dimethylamino)germanium(IV) ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99%
CAS No.: 7344-40-3
Molecular Formula: C8H24GeN4
Formula Weight: 248.90
Flash Point: 34°(93°F)
Physical Form: Liquid
Density: 1.07
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Amine-like
Melting Point: 14°
Boiling Point: 203°
UN No.: UN3129
MDL No.: MFCD01863476
Applications: Preparation of nitrides
Formel:
C₈H₂₄GeN₄ Siedepunkt: 203 °C Schmelzpunkt: 14 °C Dichte: 1,07 Flammpunkt: 34 °C (93 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863476 CAS-Nummer: 7344-40-3 UN: 3129 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...