Methylenblau 1% (w/v) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
Synonyme:
3,7-Bis(dimethylamino)phenothiaziniumchlorid, N,N,N′,N′-Tetramethylthioninchlorid, Methylthioniniumchlorid
42771.AE
42771.AP
42771.AEEA
31
EUR
42771.AE
42771.AP
Methylenblau 1% (w/v) in wässriger Lösung
Methylenblau
Molecular Formula: C16H18ClN3S
Formula Weight: 319.86
Physical Form: Liquid
MDL No.: MFCD00012111
Formel:
C₁₆H₁₈ClN₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150008 CAS-Nummer: 61-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...