Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

427751000 427758000
427751000.EA 140 EUR
427751000. 427758000.
Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal®
Isopropenylmagnesium bromide solution

Appearance: Yellow to grey to brown Solution

Formel: C₃H₅BrMg
Siedepunkt: 67 °C (760 mmHg)
Dichte: 0,927 g/cm³
Flammpunkt: 2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192240
CAS-Nummer: 13291-18-4
UN: 3399
ADR: 4.3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR