Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
427751000
427758000
427751000.EA
140
EUR
427751000.
427758000.
Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal®
Formel:
C₃H₅BrMg Siedepunkt: 67 °C (760 mmHg) Dichte: 0,927 g/cm³ Flammpunkt: 2 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192240 CAS-Nummer: 13291-18-4 UN: 3399 ADR: 4.3,I |
Spezifikation Testergebnisse
Appearance | Yellow to grey to brown Solution |
Titration Active base | 0.45 to 0.60 M |
Melting point | 37 to 41 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...