Kieselgel amorph S.A. 105-145 m²/g, gedampft, oberflächenbehandelt

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Silica gel amorphous, Silicon dioxide amorphous, Silicium dioxide amorphous

42783.18 42783.30 42783.A1
42783.18EA 24.3 EUR
42783.18 42783.30 42783.A1
Kieselgel amorph S.A. 105-145 m²/g, gedampft, oberflächenbehandelt
Kieselgel amorph

CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: -325 mesh powder Density: 2.2 Appearance: White Odour: Odourless MDL No.: MFCD00011232


Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity

Formel: SiO₂
Molecular Weight: 60.08 g/mol
Schmelzpunkt: 2230
Dichte: 2.2
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 7631-86-9
EINECS: 231-545-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR