Thymolblau Natriumsalz ACS
Lieferant: Thermo Scientific
Synonyme:
Thymolsulfonphthalein Natriumsalz, Natriumhydrogen-4,4'-(3H-2,1-benzoxathiol-3-yliden)bis[5-methyl-2-(1-methylethyl)phenolat]-S,S-dioxid, Thymolblau wasserlöslich
42785.09
42785.09EA
51.2
EUR
42785.09
Thymolblau Natriumsalz ACS
Thymolblau Natriumsalz
CAS No.: 62625-21-2
Molecular Formula: C27H29NaO5S
Formula Weight: 488.58
Physical Form: Crystalline
Melting Point: ca 284° dec.
Solubility: Soluble in water
Beilstein Reference: 3861515
MDL No.: MFCD00151093
Water soluble pH indicator
Formel:
C₂₇H₂₉NaO₅S Molecular Weight: 488,58 g/mol Schmelzpunkt: ∼ 284 °C (Zersetzung) Dichte: 1,19 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151093 CAS-Nummer: 62625-21-2 EINECS: 263-650-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...