Tetrachlorgold(III)säure Hydrat ≥99,999% (Metall-Basis) Au ≥ 49.0%, Premion®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Tetrachlorgoldsäure Hydrat, Hydrogentetrachloroaurat(III) Hydrat

42803.06 42803.03
42803.06EA 816 EUR
42803.06 42803.03
Tetrachlorgold(III)säure Hydrat ≥99,999% (Metall-Basis) Au ≥ 49.0%, Premion®
Tetrachlorgold(III)säure Hydrat

Purity: 99.999% CAS No.: 27988-77-8 Molecular Formula: HAuCl4.xH2O Formula Weight: 339.79(anhy) Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: Yellow-orange Odour: Odourless UN No.: 3260 MDL No.: MFCD00149903

Formel: HAuCl₄·H₂O
Molecular Weight: 357.8 g/mol
Dichte: 1.169 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149903
CAS-Nummer: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR