Cobalt(II)isopropoxid

Lieferant: Thermo Fisher Scientific
Achtung


Unter Argon in Ampullen verpackt

42810.03 42810.06
42810.03EA 90.3 EUR
42810.03 42810.06
Cobalt(II)isopropoxid
Cobalt(II)isopropoxid

CAS No.: 60406-95-3 Molecular Formula: Co[OCH(CH3)2]2 Formula Weight: 178.11 Physical Form: Powder, Ampouled under argon Sensitivity: Moisture Sensitive MDL No.: MFCD00799075

Formel: C₆H₁₄CoO₂
Molecular Weight: 178.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00799075
CAS-Nummer: 60406-95-3
UN: 3181
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR