Cobalt(II)isopropoxid
Lieferant: Thermo Fisher Scientific
42810.03
42810.06
42810.03EA
92.6
EUR
42810.03
42810.06
Cobalt(II)isopropoxid
Cobalt(II)isopropoxid
CAS No.: 60406-95-3
Molecular Formula: Co[OCH(CH3)2]2
Formula Weight: 178.11
Physical Form: Powder, Ampouled under argon
Sensitivity: Moisture Sensitive
MDL No.: MFCD00799075
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...